missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-2-quinolinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10768103
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10768103 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10768103 Supplier Thermo Scientific Maybridge Supplier No. AC34762DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1571-30-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point 219°C to 223°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H6NO3
MDL Number MFCD00168972
Quantity 1 g
Synonym 8-hydroxyquinaldic acid, quinaldic acid, 8-hydroxy, 2-quinolinecarboxylic acid, 8-hydroxy, 8-hydroxy-2-quinolinecarboxylic acid, 8hc, acmc-20a4qz, 5-22-05-00272 beilstein handbook reference, 8-hydroxyquinolin-2-carboxylic acid, 8-hydroxyquinoline-2-carbaxylicacid, quinolinecarboxylic acid, 8-hydroxy
InChI Key UHBIKXOBLZWFKM-UHFFFAOYSA-M
SMILES OC1=C2N=C(C=CC2=CC=C1)C([O-])=O
IUPAC Name 8-hydroxyquinoline-2-carboxylic acid
Molecular Weight (g/mol) 188.16
PubChem CID 74079
Formula Weight 189.17
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 8-Hydroxy-2-quinolinecarboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.