missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-14-nitro-10-oxo-8,14-dihydrothebaine, TRC

Product Code. 30390338
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390338 25 mg -
30394169 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390338 Supplier Toronto Research Chemicals Supplier No. TRCH94870525MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74261-01-1
Molecular Formula C19H20N2O7
Molecular Weight (g/mol) 388.37
Synonym (5alpha,8beta)-6,7-Didehydro-4,5-epoxy-8-hydroxy-3,6-dimethoxy-17-methyl-14-nitro-morphinan-10-one
IUPAC Name (4S,4aS,5S,7aR,12bR)-5-hydroxy-7,9-dimethoxy-3-methyl-4a-nitro-2,4,5,7a-tetrahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-13-one
SMILES COC1=C[C@H](O)[C@]2([C@@H]3N(C)CC[C@]24[C@H]1Oc5c(OC)ccc(C3=O)c45)[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H20N2O7/c1-20-7-6-18-13-9-4-5-10(26-2)15(13)28-17(18)11(27-3)8-12(22)19(18,21(24)25)16(20)14(9)23/h4-5,8,12,16-17,22H,6-7H2,1-3H3/t12-,16+,17-,18-,19+/m0/s1
Formula Weight 388.1271
Chemical Name or Material 8-Hydroxy-14-nitro-10-oxo-8,14-dihydrothebaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.