missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 8-Hydroxy-1,3,6-pyrenetrisulfonic acid trisodium salt, 98%

Product Code. 10444891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10444891 5 g Glass bottle 5g
10240073 1 g Glass bottle 1g
10445005 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10444891 Supplier Thermo Scientific Chemicals Supplier No. 412180050
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6358-69-6 | C16H7Na3O10S3 | 524.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6358-69-6
Molecular Formula C16H7Na3O10S3
Molecular Weight (g/mol) 524.37
MDL Number MFCD00037575
InChI Key KXXXUIKPSVVSAW-UHFFFAOYSA-K
PubChem CID 61388
ChEBI CHEBI:52083
IUPAC Name trisodium;8-hydroxypyrene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].OC1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Green-Yellow to Yellow
Density 2.1500g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 8065
Specific Gravity 2.15
Synonym pyranine, solvent green 7, c.i. solvent green 7, japan green 204, hpts, FD&C green no. 8, japan green no. 204, pyrene 1, pyranine 120, FD&C green 8
Solubility Information Solubility in water: 300g/L (25°C).
Formula Weight 524.39
Percent Purity 98%
Grade Extra Pure
Physical Form Crystals or Powder
Chemical Name or Material 8-Hydroxy-1, 3, 6-pyrenetrisulfonic acid trisodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.