missing translation for 'onlineSavingsMsg'
Learn More

8-Formyl-4-methylumbelliferone, TRC

Product Code. 30391737
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391737 10 mg
30393177 25 mg
30408554 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391737 Supplier Toronto Research Chemicals Supplier No. TRCF70083510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14003-96-4
Molecular Formula C11 H8 O4
Molecular Weight (g/mol) 204.18
Synonym 8-Formyl-7-hydroxy-4-methylcoumarin, 4-Methyl-7-hydroxy-8-formylcoumarin, 7-Hydroxy-8-formyl-4-methylcoumarin, 8-Formyl-7-hydroxy-4-methyl-2H-1-benzopyran-2-one, 8-Formyl-7-hydroxy-4-methylcoumarin, 4μ8C
IUPAC Name 7-hydroxy-4-methyl-2-oxochromene-8-carbaldehyde
SMILES CC1=CC(=O)Oc2c(C=O)c(O)ccc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H8O4/c1-6-4-10(14)15-11-7(6)2-3-9(13)8(11)5-12/h2-5,13H,1H3
Formula Weight 204.0423
Chemical Name or Material 8-Formyl-4-methylumbelliferone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.