missing translation for 'onlineSavingsMsg'
Learn More

8-(Fmoc-amino)-3,6-dioxaoctanoic Acid, TRC

Product Code. 30397212
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397212 1 g
30403587 250 mg
30394387 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397212 Supplier Toronto Research Chemicals Supplier No. TRCF6035151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166108-71-0
Molecular Formula C21H23NO6
Molecular Weight (g/mol) 385.41
Synonym Fmoc-AEEA, 12-(9H-Fluoren-9-yl)-10-oxo-3,6,11-trioxa-9-azadodecanoic Acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic Acid, 8-(9-Fluorenylmethoxycarbonylamino)-3,6-dioxaoctanoic Acid, 9-Fluorenylmethoxycarbonyl-8-amino-3,6-dioxaoctanoic Acid
IUPAC Name 2-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]acetic acid
SMILES O=C(NCCOCCOCC(O)=O)OCC1C(C=CC=C2)=C2C3=CC=CC=C13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H23NO6/c23-20(24)14-27-12-11-26-10-9-22-21(25)28-13-19-17-7-3-1-5-15(17)16-6-2-4-8-18(16)19/h1-8,19H,9-14H2,(H,22,25)(H,23,24)
Formula Weight 385.1525
Chemical Name or Material 8-(Fmoc-amino)-3,6-dioxaoctanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.