| CAS |
82664-20-8
|
| Formula Weight |
751.4518 |
| InChI Formula |
InChI=1S/C37H66FNO13/c1-14-24-37(10,46)29(42)21(5)28(41)34(7,38)17-35(8,45)31(52-33-26(40)23(39(11)12)15-18(2)48-33)19(3)27(20(4)32(44)50-24)51-25-16-36(9,47-13)30(43)22(6)49-25/h18-27,29-31,33,40,42-43,45-46H,14-17H2,1-13H3/t18-,19+,20-,21+,22+,23+,24-,25+,26-,27+,29-,30+,31-,33+,34+,35-,36-,37-/m1/s1 |
| IUPAC Name |
(3R,4S,5S,6R,7R,9S,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-9-fluoro-7,12,13-trihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione |
| Molecular Formula |
C37 H66 F N O13 |
| Molecular Weight (g/mol) |
751.92 |
| Percent Purity |
>95% |
| SMILES |
CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@](C)(F)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O |
| Synonym |
Erythromycin, 8-fluoro- (9CI), Oxacyclotetradecane, erythromycin deriv., (8S)-8-Fluoroerythromycin A, 8-Fluoroerythromycin, 8-Fluoroerythromycin A, Antibiotic P 80206, Flurithromycin, P 0501A, P 80206 |
| Chemical Name or Material |
8-Fluoro Erythromycin |
| Grade |
HPLC |
|
Show More
Show Less
|
|