missing translation for 'onlineSavingsMsg'
Learn More

8-Fluoro-6-methoxy Moxifloxacin Dihydrochloride, TRC

Product Code. 30399318
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399318 1 mg
30397484 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399318 Supplier Toronto Research Chemicals Supplier No. TRCF5933001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21H26Cl2FN3O4
Molecular Weight (g/mol) 474.35
Synonym 1-Cyclopropyl-8-fluoro-1,4-dihydro-6-methoxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid Dihydrochloride, Moxifloxacin Imp. D (EP/BP) as diHCl salt, Moxifloaxacin EP Impurity D
SMILES O=C1C2=CC(OC)=C(N3C[C@@](CCCN4)([H])[C@@]4([H])C3)C(F)=C2N(C5CC5)C=C1C(O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H24FN3O4/c1-29-16-7-13-18(25(12-4-5-12)9-14(20(13)26)21(27)28)17(22)19(16)24-8-11-3-2-6-23-15(11)10-24/h7,9,11-12,15,23H,2-6,8,10H2,1H3,(H,27,28)/t11-,15+/m0/s1
Chemical Name or Material 8-Fluoro-6-methoxy Moxifloxacin Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.