missing translation for 'onlineSavingsMsg'
Learn More

8-Ethoxyquinoline-5-sulfonic acid sodium salt hydrate, 97%, Thermo Scientific™

Product Code. 11492648
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492648 50 g 50g
11482648 10 g 10g
11402658 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492648 Supplier Thermo Scientific Alfa Aesar Supplier No. A16668.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80789-76-0
Molecular Formula C11H10NNaO4S
Molecular Weight (g/mol) 275.25
MDL Number MFCD00149478
InChI Key KGBPIIYKHSGTAJ-UHFFFAOYSA-M
Synonym sodium 8-ethoxyquinoline-5-sulfonate hydrate, 8-ethoxyquinoline-5-sulfonic acid sodium salt hemihydroate, actinoquinol sodium salt, 8-ethoxyquinoline-5-sulfonic acid sodium salt hydrate, 8-ethoxy-5-quinolinesulfonic acid sodium salt monohydrate, acmc-2097kw, sodium 8-ethoxyquinoline-5-sulfonate monohydrate, 8-ethoxyquinoline-5-sulfonic acid sodium salt hemihydrate
PubChem CID 51063116
IUPAC Name sodium;8-ethoxyquinoline-5-sulfonate;hydrate
SMILES [Na+].CCOC1=CC=C(C2=CC=CN=C12)S([O-])(=O)=O

Specifications

Melting Point >280°C
Quantity 50 g
Sensitivity Hygroscopic
Formula Weight 275.26 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 8-Ethoxyquinoline-5-sulfonic acid sodium salt hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.