missing translation for 'onlineSavingsMsg'
Learn More

8-Ethoxyquinoline-5-sulfonic acid monohydrate, 99%

Product Code. 11458557
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11458557

Brand: Thermo Scientific Alfa Aesar A18622.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15301-40-3 | C11H11NO4S | 253.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15301-40-3
Molecular Formula C11H11NO4S
Molecular Weight (g/mol) 253.27
MDL Number MFCD00065059
InChI Key YAMVZYRZAMBCED-UHFFFAOYSA-N
Synonym actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol
PubChem CID 23674
IUPAC Name 8-ethoxyquinoline-5-sulfonic acid
SMILES CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O

Specifications

Melting Point ∼296°C (decomposition)
Quantity 10 g
Beilstein 225878
Formula Weight 271.29 (253.28 Anhydrous)
Percent Purity 99%
Chemical Name or Material 8-Ethoxyquinoline-5-sulfonic acid monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.