missing translation for 'onlineSavingsMsg'
Learn More

8-Dimethylamino-2,3-Benzophenoxazine Hemi (Zinc Chloride) Salt, MP Biomedicals™

Product Code. 11471191 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471191 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471191 Supplier MP Biomedicals Supplier No. 0215778805
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7057-57-0
Molecular Formula C18H15ClN2O
Quantity 5 g
Synonym basic blue 6, meldola's blue, phenylene blue, unii-cp2st8ed1o, c.i. basic blue 6, cp2st8ed1o, 9-dimethylamino benzo a phenoxazin-7-ium chloride, benzo a phenoxazin-7-ium, 9-dimethylamino-, chloride, 8-dimethylamino-2,3-benzophenoxazine hemi zinc chloride salt, n,n-dimethyl-10-oxa-5-azatetraphen-8-iminium chloride
InChI Key HWYNRVXFYFQSID-UHFFFAOYSA-M
SMILES C[N+](=C1C=CC2=NC3=C(C=CC4=CC=CC=C43)OC2=C1)C.[Cl-]
IUPAC Name benzo[a]phenoxazin-9-ylidene(dimethyl)azanium;chloride
Molecular Weight (g/mol) 310.781
PubChem CID 101929
Formula Weight 378.9
Physical Form Solid
Chemical Name or Material 8-Dimethylamino-2, 3-Benzophenoxazine Hemi (Zinc Chloride) Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.