missing translation for 'onlineSavingsMsg'
Learn More

8-Demethoxy-8-fluoro Gatifloxacin, TRC

Product Code. 30365772
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365772 10 mg 10mg
30351601 5 mg 5mg
30355445 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365772 Supplier Toronto Research Chemicals Supplier No. TRCD23080010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 103460-89-5
Molecular Formula C18 H19 F2 N3 O3
Molecular Weight (g/mol) 363.36
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-, 1-Cyclopropyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, 1-Cyclopropyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid, AMQ 2, CP 105532, 8-Demethoxy-8-fluoro Gatifloxacin, 8-Demethoxy-8-fluorogatifloxacin
IUPAC Name 1-cyclopropyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
SMILES CC1CN(CCN1)c2c(F)cc3C(=O)C(=CN(C4CC4)c3c2F)C(=O)O

Tekniske data

Quantity 10 mg
InChI Formula InChI=1S/C18H19F2N3O3/c1-9-7-22(5-4-21-9)16-13(19)6-11-15(14(16)20)23(10-2-3-10)8-12(17(11)24)18(25)26/h6,8-10,21H,2-5,7H2,1H3,(H,25,26)
Formula Weight 363.1394
Chemical Name or Material 8-Demethoxy-8-fluoro Gatifloxacin
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.