missing translation for 'onlineSavingsMsg'
Learn More

8 Component Mix in DMSO, SPEX CertiPrep™

Product Code. 16390498 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16390498 1 mL 1mL
16310508 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16390498 Supplier SPEX Certiprep Supplier No. USPRSC2B
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Chemical Name or Material 8 Component Mix in DMSO
Name Note USP 467 Standard
Packaging Glass Ampoule
CAS 127-19-5, 68-12-2, 107-21-1, 872-50-4, 75-12-7
Explosion Limit 1.8% (volume); 63% (volume)
Ignition Point 270°C
Vapor Pressure 2.5 hPA (20°C)
Methods USP 467: Residual Solvents/ Organic Volatile Impurities
Analyte or Component Names N,N-Dimethylacetamide; N,N-Dimethylformamide; Ethylene glycol; 1-Methyl-2-pyrrolidinone; Formamide; more
Density 1.1g/cm3
Physical Form Fluid
Element N,N-Dimethylacetamide, N,N-Dimethylformamide, Ethylene Glycol, 1-Methyl-2-pyrrolidinone, Formamide
Solvent or Matrix Dimethyl Sulfoxide
Quantity 1 mL
Grade USP
Show More Show Less
compliance-icons
Product Identifier
  • Stock VOA
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: dimethyl sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.