missing translation for 'onlineSavingsMsg'
Learn More

8-chloronaphthalene-1-sulfonic acid, 97%, Thermo Scientific™

Product Code. 10011664
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10011664 25 g 25g
10784183 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10011664 Supplier Thermo Scientific Maybridge Supplier No. BTB06262EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 145-74-4
Molecular Formula C10H7ClO3S
Molecular Weight (g/mol) 242.673
MDL Number MFCD00052379
InChI Key OPFNCYFEIBUZHU-UHFFFAOYSA-N
Synonym 8-chloronaphthalene-1-sulphonic acid, 8-chloro-1-naphthalenesulfonic acid, 1-naphthalenesulfonicacid, 8-chloro, sodium 8-chloronaphthalene-1-sulphonate, maybridge1_001443, 1-naphthalenesulfonic acid, 8-chloro
PubChem CID 67353
IUPAC Name 8-chloronaphthalene-1-sulfonic acid
SMILES C1=CC2=C(C(=C1)S(=O)(=O)O)C(=CC=C2)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 242.68
Percent Purity 97%
Chemical Name or Material 8-chloronaphthalene-1-sulfonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.