missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Mikromol™

Product Code. 17931606
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17931606 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17931606 Supplier Mikromol Supplier No. MM0257.250025
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

8-Chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
CAS 1246819-22-6
Analyte or Component Names 8-Chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Shipping Condition Room Temperature
Molecular Formula C19 H18 Cl N3 O
Certifications/Compliance ISO 9001
Synonym 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 8-chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-, 8-Chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, N-Nitroso desloratadine, N-Nitrosodesloratadine
InChI Formula InChI=1S/C19H18ClN3O/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-21-19(14)18(17)13-7-10-23(22-24)11-8-13/h1-2,5-6,9,12H,3-4,7-8,10-11H2
SMILES Clc1ccc2C(=C3CCN(CC3)N=O)c4ncccc4CCc2c1
IUPAC Name 8-chloro-11-(1-nitrosopiperidin-4-ylidene)-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
Molecular Weight (g/mol) 339.8187
Quantity 25 mg
Formula Weight 339.1138 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.