missing translation for 'onlineSavingsMsg'
Learn More

8-chloro-2-methylquinoline, 97%, Thermo Scientific™

Product Code. 10576324
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10576324 25 g 25g
10213522 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10576324 Supplier Thermo Scientific Maybridge Supplier No. RJC01118EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3033-82-7
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.631
MDL Number MFCD00051764
InChI Key VVLYDFPOGMTMFJ-UHFFFAOYSA-N
Synonym 8-chloroquinaldine, quinaldine, 8-chloro, quinoline, 8-chloro-2-methyl, 8-chloro-2-methyl-quinoline, 2-methyl-8-chloroquinoline, pubchem5852, acmc-1aehk, maybridge1_006766, 8-chloranyl-2-methyl-quinoline, 8-chloro-2-methylquinoline
PubChem CID 221113
IUPAC Name 8-chloro-2-methylquinoline
SMILES CC1=NC2=C(C=CC=C2Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 177.63
Percent Purity 97%
Chemical Name or Material 8-Chloro-2-methylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.