missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-11-(piperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine (Descarboethoxyloratadine; Desloratadine), Mikromol™

Product Code. 17990956
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17990956 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17990956 Supplier Mikromol Supplier No. MM0257.010250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

8-Chloro-11-(piperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine (Descarboethoxyloratadine; Desloratadine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desloratadine
CAS 100643-71-8
Analyte or Component Names Desloratadine
Shipping Condition Room Temperature
Molecular Formula C19 H19 Cl N2
Certifications/Compliance ISO 17034
Synonym 8-Chloro-6,11-dihydro-11-(4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Aerius, Allex, Azomyr, Clarinex, Descarboethoxyloratadine, NSC 675447, Neoclarityn, Opulis, Sch 34117, USP Loratadine Related Compound A, 8-Chloro-11-(4-piperidylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Delopedil, Descarboethoxyloratidine, Desler, Desloratadine, Eurodesa, Sedno, 8-Chloro-6,11-dihydro-11-(4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Aerius, Allex, Azomyr, Clarinex, Descarboethoxyloratadine, Neoclarityn, Opulis, Sch 34117, USP Loratadine Related Compound A, Loratadine EP Impurity D, Loratadine USP Related Compound A, Loratadine Related Compound A, Loratadine Impurity D, Rupatadine Fumarate Impurity B
InChI Formula InChI=1S/C19H19ClN2/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-22-19(14)18(17)13-7-10-21-11-8-13/h1-2,5-6,9,12,21H,3-4,7-8,10-11H2
SMILES Clc1ccc2C(=C3CCNCC3)c4ncccc4CCc2c1
IUPAC Name 8-chloro-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
Molecular Weight (g/mol) 310.82
Quantity 250 mg
Formula Weight 310.1237 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.