missing translation for 'onlineSavingsMsg'
Learn More

8-Carboxynaphthalene-1-carboxamide, tech. 85%, Thermo Scientific™

Product Code. 11343276
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11343276

Brand: Thermo Scientific Alfa Aesar L12628.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5811-88-1
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD00052020
InChI Key GRVIMBYDMARKEZ-UHFFFAOYSA-N
Synonym 8-aminocarbonyl-1-naphthoic acid, 8-carboxynaphthalene-1-carboxamide, 8-carbamoyl-1-naphthoic acid, 8-carbamoylnaphthalenecarboxylic acid, maybridge1_003186, acmc-1am92, 1,8-naphthalenedicarboxylic imide, 8-carboxynaphyhalene-1-carboxamide, 8-carbamoyl-1-naphthalenecarboxylic acid, 8-aminocarbonylnaphthalene-1-carboxylic acid
PubChem CID 2777172
IUPAC Name 8-carbamoylnaphthalene-1-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)N)C(=CC=C2)C(=O)O

Specifications

Melting Point 265°C to 270°C
Quantity 25 g
Beilstein 2213597
Formula Weight 215.21
Percent Purity 85%
Grade Technical
Chemical Name or Material 8-Carboxynaphthalene-1-carboxamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.