missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoxanthine, TRC

Product Code. 30331581
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331581 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331581 Supplier Toronto Research Chemicals Supplier No. TRCB6891801G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 8-Bromoxanthine
CAS 10357-68-3
Molecular Formula C5H3BrN4O2
Quantity 1 g
Synonym 8-Bromo-3,7-dihydro-1H-purine-2,6-dione, 8-Bromo-xanthine, 8-Bromopurine-2,6-diol, NSC 24131
InChI Formula InChI=1S/C5H3BrN4O2/c6-4-7-1-2(8-4)9-5(12)10-3(1)11/h(H3,7,8,9,10,11,12)
SMILES Brc1nc2NC(=O)NC(=O)c2[nH]1
IUPAC Name 8-bromo-3,7-dihydropurine-2,6-dione
Molecular Weight (g/mol) 231.01
Formula Weight 229.9439
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.