missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoquinoline, 98%, Thermo Scientific Chemicals

Product Code. 11405750
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405750 1 g 1g
15483327 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405750 Supplier Thermo Scientific Alfa Aesar Supplier No. H25961.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16567-18-3 | Molecular Formula: C9H6BrN | Molecular Weight: 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16567-18-3
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00191859
InChI Key PIWNKSHCLTZKSZ-UHFFFAOYSA-N
Synonym quinoline, 8-bromo, 8-bromo-quinoline, 8-bromoquinoline, pubchem5816, 8-bromquinoline, acmc-209dt3, ksc181q5t, buttpark 33\04-57
PubChem CID 140109
IUPAC Name 8-bromoquinoline
SMILES BrC1=C2N=CC=CC2=CC=C1

Specifications

Density 1.594
Boiling Point 302°C to 304°C
Flash Point >110°C (230°F)
Refractive Index 1.6725
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 8-Bromoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.