missing translation for 'onlineSavingsMsg'
Learn More

8-Bromooctanoic acid, 95%, Thermo Scientific Chemicals

Product Code. 10713413
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17696-11-6
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00004430
InChI Key BKJFDZSBZWHRNH-UHFFFAOYSA-N
Synonym 8-bromocaprylic acid, 8-bromo octanoic acid, octanoic acid, 8-bromo, 8-bromo-n-octanoic acid, omega-bromocaprylic acid, 8-bromo-octanoic acid, octanoicacid, 8-bromo, acmc-1bo24, 8-bromooctanoic acid, ksc180g8h
PubChem CID 548275
IUPAC Name 8-bromooctanoic acid
SMILES C(CCCC(=O)O)CCCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 36.0°C to 41.0°C
Color White to Cream
Boiling Point 147.0°C to 150.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 02, IV, 1000
Formula Weight 223.11
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromooctanoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 8-Bromooctanoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'