missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoimidazo[1,2-a]pyridine, 97%, Alfa Aesar™

Product Code. 15436717
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436717 250 mg 250mg
15426717 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436717 Supplier Thermo Scientific Alfa Aesar Supplier No. H33841.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 850349-02-9
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD08706060
InChI Key GCCVNNZARPFXES-UHFFFAOYSA-N
Synonym 8-bromoimidazo 1,2-a pyridine, 8-bromo-imidazo 1,2-a pyridine, 8-bromoh-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine,8-bromo, imidazo 1,2-a pyridine, 8-bromo
PubChem CID 17749933
IUPAC Name 8-bromoimidazo[1,2-a]pyridine
SMILES C1=CN2C=CN=C2C(=C1)Br

Specifications

Melting Point 72°C to 74°C
Quantity 250 mg
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 8-Bromoimidazo[1,2-a]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.