missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenosine, 98%

Product Code. 11332216
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332216 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11332216 Supplier Thermo Scientific Alfa Aesar Supplier No. L03544.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2946-39-6 | C10H12BrN5O4 | 346.14 g/mol

8-Bromoadenosine is an adenosine binding inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromoadenosine is an adenosine binding inhibitor.

Solubility
Soluble in DMSO, and DMF. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2946-39-6
Molecular Formula C10H12BrN5O4
Molecular Weight (g/mol) 346.14
MDL Number MFCD00005733
InChI Key VJUPMOPLUQHMLE-SBTUYZBDNA-N
Synonym 8-bromoadenosine, bromoadenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, adenosine, 8-bromo, 6-amino-8-bromopurine riboside, 8-bromo-adenosine, 8-bromoadenine-9-beta-d-ribofuranoside, 8-bromo adenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 4s,2r,3r,5r-2-6-amino-8-bromopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol
PubChem CID 96544
ChEBI CHEBI:2312
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-8-bromopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=C(Br)N([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Melting Point ∽211°C (decomposition)
Quantity 250 mg
Beilstein 627737
Sensitivity Air sensitive
Solubility Information Soluble in DMSO,and DMF. Insoluble in water.
Optical Rotation −33° (c=1 in 1N HCl)
Formula Weight 346.15
Percent Purity 98%
Chemical Name or Material 8-Bromoadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.