missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenosine, 98%

Product Code. 10317380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317380 5 g Glass bottle 5g
10520424 250 mg Glass bottle 250mg
10685012 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10317380 Supplier Thermo Scientific Acros Supplier No. 155780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2946-39-6 | C10H12BrN5O4 | 346.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2946-39-6
Molecular Formula C10H12BrN5O4
Molecular Weight (g/mol) 346.14
MDL Number MFCD00005733
InChI Key VJUPMOPLUQHMLE-SBTUYZBDNA-N
Synonym 8-bromoadenosine, bromoadenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, adenosine, 8-bromo, 6-amino-8-bromopurine riboside, 8-bromo-adenosine, 8-bromoadenine-9-beta-d-ribofuranoside, 8-bromo adenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 4s,2r,3r,5r-2-6-amino-8-bromopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol
PubChem CID 96544
ChEBI CHEBI:2312
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-8-bromopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=C(Br)N([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 210°C to 212°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation -54.5
Formula Weight 346.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromoadenosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.