missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenine

Product Code. 15436979
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436979 100 mg 100mg
15446979 500 mg 500mg
15426979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15436979 Supplier Thermo Scientific Alfa Aesar Supplier No. J65582.MC
  • ONLINE EXCLUSIVE PRICE
    £62.30
    £56.65 / 100mg
    Save £5.65
    9% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6974-78-3 | C5H4BrN5 | 214.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6974-78-3
Molecular Formula C5H4BrN5
Molecular Weight (g/mol) 214.03
MDL Number MFCD00082518
InChI Key FVXHPCVBOXMRJP-UHFFFAOYSA-N
Synonym 8-bromoadenine, 8-bromo-9h-purin-6-amine, 6-amino-8-bromopurine, 8-br-adenine, 8-bromo-1h-adenine, 8-bromo-1h-purin-6-amine, 8-bromo-9h-purin-6-ylamine, 8-bromopurine-6-ylamine, chembl65747, 9h-purin-6-amine, 8-bromo
PubChem CID 81457
IUPAC Name 8-bromo-7H-purin-6-amine
SMILES NC1=C2NC(Br)=NC2=NC=N1

Specifications

Melting Point >250°C
Color Brown to White
Quantity 100 mg
Formula Weight 214.03
Physical Form Powder
Chemical Name or Material 8-Bromoadenine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.