missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 8-Bromo-cGMP, sodium salt
GREENER_CHOICE

Product Code. 18095822
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18095822 50 mg 50mg
18094032 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095822 Supplier Tocris Bioscience™ Supplier No. 1089/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

cGMP analog; activates PKG

Membrane permeable cGMP analog that activates protein kinase G (PKG). 4.3-fold more potent than cGMP in activating PKG1α and promotes relaxation of tracheal and vascular smooth muscle tissue in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51116-01-9
Molecular Formula C10H11BrN5O7P

Specifications

Quantity 50 mg
Formula Weight 446.09
Purity >98%
Molecular Weight (g/mol) 424.10
Chemical Name or Material 8-Bromo-cGMP, sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.