missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 8-Bromo-cAMP

Product Code. 17120252
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17120252 100 mg 100mg
17110252 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17120252 Supplier STEMCELL Technologies Supplier No. 73604
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

cAMP pathway activator; Activates cAMP-dependent kinase

  • 8-Bromo-cAMP is a membrane-permeable cAMP derivative
  • It can activate cAMP-dependent protein kinase, with long-acting effects due to its resistance to cAMP phosphodiesterase (Schwede et al.) It can be used to study calcium-mediated pathways (IC50 = 0.84 mM; Xaus et al.) REPROGRAMMING
  • Improves the reprogramming efficiency of human neonatal foreskin fibroblast (HFF1) cells, in combination with vaLproic acid (Wang & Adjaye)
  • Inhibits M-CSF-dependent proliferation of macrophages (Xaus et al.)
  • Protects neutrophils against TNF-α-induced apoptosis (Krakstad)
  • Induces a proliferative response in an IL-3-dependent leukemic cell line (Barge et al.)
  • Induces membrane depolarization in pancreatic cancer cell lines (Sorio et al.)

Specifications

CAS 23583-48-4
Synonym 8-BrcAMP; 8-Bromoadenosine 3’,5’-cyclic monophosphate; NSC 171719
Quantity 100 mg
Molecular Formula C10H11BrN5O6P
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.