missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies 8-Bromo-cAMP
Description
- 8-Bromo-cAMP is a membrane-permeable cAMP derivative
- It can activate cAMP-dependent protein kinase, with long-acting effects due to its resistance to cAMP phosphodiesterase (Schwede et al.) It can be used to study calcium-mediated pathways (IC50 = 0.84 mM; Xaus et al.) REPROGRAMMING
- Improves the reprogramming efficiency of human neonatal foreskin fibroblast (HFF1) cells, in combination with vaLproic acid (Wang & Adjaye)
- Inhibits M-CSF-dependent proliferation of macrophages (Xaus et al.)
- Protects neutrophils against TNF-α-induced apoptosis (Krakstad)
- Induces a proliferative response in an IL-3-dependent leukemic cell line (Barge et al.)
- Induces membrane depolarization in pancreatic cancer cell lines (Sorio et al.)
Specifications
Specifications
| CAS | 23583-48-4 |
| Synonym | 8-BrcAMP; 8-Bromoadenosine 3',5'-cyclic monophosphate; NSC 171719 |
| Quantity | 10 mg |
| Molecular Formula | C10H11BrN5O6P |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?