missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo Adenosine, TRC

Product Code. 30331621
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331621 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331621 Supplier Toronto Research Chemicals Supplier No. TRCB6793751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Bromo Adenosine
CAS 2946-39-6
Molecular Formula C10 H12 Br N5 O4
Quantity 1 g
InChI Formula InChI=1S/C10H12BrN5O4/c11-10-15-4-7(12)13-2-14-8(4)16(10)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,13,14)/t3-,5-,6-,9-/m1/s1
SMILES Nc1ncnc2c1nc(Br)n2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-8-bromopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 346.14
Formula Weight 345.0073
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.