missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-naphthoic acid, Technical Grade, Maybridge™

Product Code. p-4509713
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10039161

Brand: Thermo Scientific Maybridge BTB12326DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1729-99-3
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
InChI Key DMEZDDHJCUHENA-UHFFFAOYSA-N
Synonym 8-bromo-1-naphthoic acid, 1-naphthalenecarboxylic acid, 8-bromo, maybridge1_002138, 8-bromo-1-carboxynaphthalene, 8-bromo-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 8-bromo, 8-bromo-naphthalene-1-carboxylic acid
PubChem CID 72873
IUPAC Name 8-bromonaphthalene-1-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)C(=CC=C2)Br

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 251.08
Percent Purity 95%
Chemical Name or Material 8-Bromo-1-naphthoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.