missing translation for 'onlineSavingsMsg'
Learn More

8-Azahypoxanthine, 98%

Product Code. 10194260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10194260 1 g 1g
12181050 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10194260 Supplier Thermo Scientific Acros Supplier No. 202590010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2683-90-1 | C4H3N5O | 137.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2683-90-1
Molecular Formula C4H3N5O
Molecular Weight (g/mol) 137.10
MDL Number MFCD00005804
InChI Key OEEYCNOOAHGFHL-UHFFFAOYSA-N
Synonym 8-azahypoxanthine, azahypoxanthine, 3h-1,2,3 triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-1,2,3,4,6-pentaazaindene, 7h-1,2,3-triazolo 4,5-d pyrimidin-7-one, 1,4-dihydro, 7-hydroxy-v-triazolo d pyrimidine, 1h-1,2,3-triazolo 4,5-d pyrimidin-7-ol, v-triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-3h-1,2,3 triazolo 4,5-d pyrimidine
PubChem CID 75895
IUPAC Name 2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one
SMILES O=C1N=CN=C2NNN=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 26, IV, 4130
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution at ph >9
Formula Weight 137.1
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 8-Azahypoxanthine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 8-Azahypoxanthine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.