missing translation for 'onlineSavingsMsg'
Learn More

8-AnilinonapHthalene-1-Sulfonic Acid Ammonium Salt, Pure, Fisher Chemical

Product Code. 10539950 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10539950 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10539950 Supplier Fisher Chemical Supplier No. A/7486/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 243°C
CAS 28836-03-5
Color Green
Physical Form Solid
Quantity 250 g
Molecular Formula C16H16N2O3S
MDL Number 12560
InChI Key IPBNQYLKHUNLQE-UHFFFAOYSA-N
SMILES [NH4+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
IUPAC Name ammonium 8-(phenylamino)naphthalene-1-sulfonate
Molecular Weight (g/mol) 316.38
Formula Weight 316.38g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • 8-Anilinonaphthalene-1-sulfonic acid ammonium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.