missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Anilino-1-naphthalenesulfonic Acid, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 8-Anilino-1-naphthalenesulfonic Acid |
| CAS | 82-76-8 |
| Molecular Formula | C16H13NO3S |
| Quantity | 5 g |
| Synonym | 1-(Phenylamino)-8-naphthalenesulfonic Acid, 1-Anilino-8-naphthalenesulfonic Acid, 8-(Phenylamino)-1-naphthalenesulfonic Acid, 8-Anilino-1-naphthalenesulfonate, ANS, ANS (Fluorescent Probe), NSC 1746, Phenyl Peri Acid |
| InChI Formula | InChI=1S/C16H13NO3S/c18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13/h1-11,17H,(H,18,19,20) |
| SMILES | OS(=O)(=O)c1cccc2cccc(Nc3ccccc3)c12 |
| IUPAC Name | 8-anilinonaphthalene-1-sulfonic acid |
| Molecular Weight (g/mol) | 299.34 |
| Formula Weight | 299.0616 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction