missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Anilino-1-naphthalene sulfonic acid, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
Specifications
Specifications
| Color | Gray to Gray-Green |
| Assay Percent Range | 0.983 |
| Quantity | 1 g |
| Formula Weight | 299.35 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 8-Anilino-1-naphthalenesulfonic acid, For HPLC analysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction