missing translation for 'onlineSavingsMsg'
Learn More

8-Aminoquinoline, 98+%

Product Code. 11452768
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452768 25 g 25g
11432768 1 g 1g
11442768 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452768 Supplier Thermo Scientific Alfa Aesar Supplier No. A16752.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 578-66-5 | C9H8N2 | 144.177 g/mol

8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

Solubility
Soluble in water.

Notes
Air Sensitive. Store away from air under dry inert gas. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578-66-5
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006809
InChI Key WREVVZMUNPAPOV-UHFFFAOYSA-N
Synonym 8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
PubChem CID 11359
IUPAC Name quinolin-8-amine
SMILES C1=CC2=C(C(=C1)N)N=CC=C2

Specifications

Melting Point 63°C to 67°C
Boiling Point 174°C (26 mmHg)
Quantity 25 g
Beilstein 114474
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 144.18
Percent Purity ≥98%
Chemical Name or Material 8-Aminoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.