missing translation for 'onlineSavingsMsg'
Learn More

8-Aminonaphthalene-2-sulfonic acid, 95%

Product Code. 15426829
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15426829

Brand: Thermo Scientific Alfa Aesar J65304.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-28-8 | C10H9NO3S | 223.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-28-8
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00004032
InChI Key QEZZCWMQXHXAFG-UHFFFAOYSA-N
Synonym 8-amino-2-naphthalenesulfonic acid, 1,7-cleve's acid, 1-naphthylamine-7-sulfonic acid, 2-naphthalenesulfonic acid, 8-amino, cleve's theta-acid, 1-amino-7-sulfonaphthalene, 1-aminonaphthalene-7-sulfonic acid, 8-naphthylamine-2-sulfonic acid, 1-amino-7-naphthalenesulfonic acid, 1-amino-naphthalene-7-sulphonic acid
PubChem CID 8386
IUPAC Name 8-aminonaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=C(C=C2)S(=O)(=O)O)C(=C1)N

Specifications

Melting Point >300°C
Color Gray to Yellow
Quantity 100 g
UN Number UN2585
Beilstein 2115629
Merck Index 142,351
Formula Weight 223.25
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 8-Aminonaphthalene-2-sulfonic acid
compliance-icons
Product Identifier
  • 8-Aminonaphthalene-2-sulphonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 8-Aminonaphthalene-2-sulphonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.