missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech., Thermo Scientific Chemicals
£14.00 - £331.00
Chemical Identifiers
CAS | 5460-09-3 |
---|---|
Molecular Formula | C10H8NNaO7S2·H2O |
Molecular Weight (g/mol) | 359.32 |
MDL Number | MFCD00150460 |
InChI Key | QPILZZVXGUNELN-UHFFFAOYSA-M |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10780282
|
Thermo Scientific Acros
400840050 |
5 g | Glass bottle |
£14.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10761704
|
Thermo Scientific Acros
400841000 |
100 g | Plastic bottle |
£68.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10218803
|
Thermo Scientific Acros
400845000 |
500 g | Plastic Bottle |
£331.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5460-09-3 | |
359.32 | |
QPILZZVXGUNELN-UHFFFAOYSA-M |
C10H8NNaO7S2·H2O | |
MFCD00150460 |
Specifications
5460-09-3 | |
100.0 | |
4 % to 6 % (1 g, 150°C, 4 hrs) | |
75% min. (HPLC) | |
MFCD00150460 | |
Solubility in water: slightly soluble. | |
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)[O-])S(=O)(=O)O.[Na+] | |
359.32 | |
359.32 | |
Technical | |
8-Amino-1-naphthol-3, 6-disulfonic acid, monosodium salt monohydrate |
75.0 | |
Brown to Yellow | |
Authentic | |
C10H8NNaO7S2·H2O | |
h acid sodium, 1-amino-8-naphthol-3,6-disulfonic acid monosodiumsalt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid 2-sodium salt | |
QPILZZVXGUNELN-UHFFFAOYSA-M | |
sodium;8-amino-3,6-disulfonaphthalen-1-olate | |
102116466 | |
80% | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 226-736-4
RUO – Research Use Only