missing translation for 'onlineSavingsMsg'
Learn More

Clinisafe™ 8.5 L Clinical Waste Container

Clinisafe™ containers are made using low carbon footprint recycled material, providing a safe and sustainable solution to your clinical waste management needs.

Brand:  Clinisafe™ 51912430

Additional Details : Weight : 10.74000kg

 View more versions of this product

Product Code. 17611735

  • £133.00 / Pack of 20
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Clinisafe™ offer a sustainable clinical waste management solution into the market, that reduces your carbon footprint and helps you on the road to net zero.
  • The 8.5 Liter Clinisafe™ container is specifically designed to enable the safe disposal of non-sharps clinical waste. Clinisafe™ Clinical Waste Container range has been designed to hold anatomical and laboratory waste.
  • Features and benefits
    • Made from low carbon footprint recycled material
    • High-level leak-resistant hermetic seal
    • Temporary closure and 4 click final closure features to prevent potential spillages and overfilling
    • Carry handle for easy transportation
    • Large aperture
    • Compliant with UN3292
Specifications

Specifications

Waste Container
UK Regulations, UN3292
Gray
20/Cs.
8.5 L
Yellow
325 x 457 x 783 mm
Safe Disposal of Non-Sharps Clinical Waste
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers