missing translation for 'onlineSavingsMsg'
Learn More

8-(4-Chlorobutyl)-8-azaspiro[4.5]decane-7,9-dione, TRC

Product Code. 17992675
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992675 1 g 1g
17924515 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17992675 Supplier Toronto Research Chemicals Supplier No. TRCC3650201G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21098-11-3
Molecular Formula C13 H20 Cl N O2
Molecular Weight (g/mol) 257.76
Synonym 8-(4-Chlorobutyl)-8-azaspiro[4.5]-decane-7,9-dione, Buspirone Hydrochloride Imp. L (EP)
IUPAC Name 8-(4-chlorobutyl)-8-azaspiro[4.5]decane-7,9-dione
SMILES ClCCCCN1C(=O)CC2(CCCC2)CC1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H20ClNO2/c14-7-3-4-8-15-11(16)9-13(10-12(15)17)5-1-2-6-13/h1-10H2
Formula Weight 257.1183
Percent Purity >95%
Chemical Name or Material 8-(4-Chlorobutyl)-8-azaspiro[4.5]-decane-7,9-dione
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.