missing translation for 'onlineSavingsMsg'
Learn More

8-[2-(Diphenylmethoxy)-N-methylethylamino]-1,3-dimethylxanthine, TRC

Product Code. 30372291
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372291 25 mg -
30380855 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30372291 Supplier Toronto Research Chemicals Supplier No. TRCD49206525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33120-34-2
Molecular Formula C23 H25 N5 O3
Molecular Weight (g/mol) 419.48
Synonym 8-[(2-(Diphenylmethoxy)ethyl)(methyl)amino]-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione
IUPAC Name 8-[2-benzhydryloxyethyl(methyl)amino]-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN(CCOC(c1ccccc1)c2ccccc2)c3nc4N(C)C(=O)N(C)C(=O)c4[nH]3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H25N5O3/c1-26(22-24-18-20(25-22)27(2)23(30)28(3)21(18)29)14-15-31-19(16-10-6-4-7-11-16)17-12-8-5-9-13-17/h4-13,19H,14-15H2,1-3H3,(H,24,25)
Formula Weight 419.1957
Chemical Name or Material 8-[(2-(Diphenylmethoxy)ethyl)(methyl)amino]-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.