missing translation for 'onlineSavingsMsg'
Learn More

8-[(2-(Diphenylmethoxy)ethyl)(methyl)amino]-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, Mikromol™

Product Code. 17913686
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913686 25 mg 25mg
17923686 100 mg 100mg
30460195 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17913686 Supplier Mikromol Supplier No. MM0599.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

8-[(2-(Diphenylmethoxy)ethyl)(methyl)amino]-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-[(2-(Diphenylmethoxy)ethyl)(methyl)amino]-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione
CAS 33120-34-2
Analyte or Component Names 8-[(2-(Diphenylmethoxy)ethyl)(methyl)amino]-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione
Shipping Condition Room Temperature
Molecular Formula C23 H25 N5 O3
Certifications/Compliance ISO 17025
Synonym 8-[(2-(Diphenylmethoxy)ethyl)(methyl)amino]-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione
InChI Formula InChI=1S/C23H25N5O3/c1-26(22-24-18-20(25-22)27(2)23(30)28(3)21(18)29)14-15-31-19(16-10-6-4-7-11-16)17-12-8-5-9-13-17/h4-13,19H,14-15H2,1-3H3,(H,24,25)
SMILES CN(CCOC(c1ccccc1)c2ccccc2)c3nc4N(C)C(=O)N(C)C(=O)c4[nH]3
IUPAC Name 8-[2-benzhydryloxyethyl(methyl)amino]-1,3-dimethyl-7H-purine-2,6-dione
Molecular Weight (g/mol) 419.48
Quantity 25 mg
Formula Weight 419.1957 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.