missing translation for 'onlineSavingsMsg'
Learn More

8,14-Dihydroxy-2,2,20,20-tetramethyl-heneicosanedioic Acid, TRC

Product Code. 30376487
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376487 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376487 Supplier Toronto Research Chemicals Supplier No. TRCD4770772.5MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8,14-Dihydroxy-2,2,20,20-tetramethyl-heneicosanedioic Acid
CAS 2570179-40-5
Molecular Formula C25H48O6
Quantity 2.5 mg
Synonym Bempedoic Acid Impurity 3
InChI Formula InChI=1S/C25H48O6/c1-24(2,22(28)29)18-12-6-10-16-20(26)14-8-5-9-15-21(27)17-11-7-13-19-25(3,4)23(30)31/h20-21,26-27H,5-19H2,1-4H3,(H,28,29)(H,30,31)
SMILES O=C(O)C(C)(C)CCCCCC(O)CCCCCC(O)CCCCCC(C(=O)O)(C)C
IUPAC Name 8,14-dihydroxy-2,2,20,20-tetramethylhenicosanedioic acid
Molecular Weight (g/mol) 444.64
Formula Weight 444.35
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.