missing translation for 'onlineSavingsMsg'
Learn More

(7Z)-7-Hexadecenoic Acid (Solution in Ethanol), TRC

Product Code. 30391767
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391767 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391767 Supplier Toronto Research Chemicals Supplier No. TRCH2935405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (7Z)-7-Hexadecenoic Acid
CAS 2416-19-5
Molecular Formula C16 H30 O2
Quantity 5 mg
Synonym (Z)-7-Hexadecenoic acid, 7Z-Hexadecenoic acid, Hypogeic acid, cis-7-Hexadecenoic acid
InChI Formula InChI=1S/C16H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h9-10H,2-8,11-15H2,1H3,(H,17,18)/b10-9-
SMILES CCCCCCCC\C=C/CCCCCC(=O)O
IUPAC Name (Z)-hexadec-7-enoic acid
Molecular Weight (g/mol) 254.41
Formula Weight 254.2246
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.