missing translation for 'onlineSavingsMsg'
Learn More

(7Z)-7-Hexadecenoic Acid Methyl Ester (Solution in Ethanol), TRC

Product Code. 30388939
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388939 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388939 Supplier Toronto Research Chemicals Supplier No. TRCH6352351MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56875-67-3
Molecular Formula C17H32O2
Quantity 1 mg
Synonym (Z)-7-Hexadecenoic acid methyl ester, Methyl (Z)-7-hexadecenoate, Methyl cis-7-hexadecenoate
InChI Formula InChI=1S/C17H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h10-11H,3-9,12-16H2,1-2H3/b11-10-
SMILES CCCCCCCC\C=C/CCCCCC(=O)OC
IUPAC Name methyl (Z)-hexadec-7-enoate
Molecular Weight (g/mol) 268.43
Formula Weight 268.2402
Chemical Name or Material (7Z)-7-Hexadecenoic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.