missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 7X™ Laboratory Detergents

Product Code. 11489575 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
4 x 1 gal.
55 gal.
Unit Size:
208.19 L
4 GAL
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (English) unitSize
11489575 55 gal. Each
11459575 1 gal. Each
12740748 4 x 1 gal. 4 GAL
11479575 5 gal. Each
17194175 55 gal. 208.19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11489575 Supplier MP Biomedicals™ Supplier No. 097667098

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Safe, effective cleaning solutions for glass and plastic labware

7X cleaning solution was developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments

  • Effective and safe
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
pH 6 to 7.5
Concentration 7X
Content And Storage 15°C to 30°C
For Use With (Application) Use on Glass, Plastic, benchtop equipment and tubing
Volume (English) 55 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.