missing translation for 'onlineSavingsMsg'
Learn More

7rh, MedChemExpress

Product Code. 16492648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492648 5 mg 5mg
16480449 10 mg 10mg
16404116 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492648 Supplier Medchem Express Supplier No. HYU004445mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 7rh (DDR1-IN-2) is a potent inhibitor of discoidin domain receptor 1 (DDR1), with an IC50 of 13.1 nM, and also less potently inhibits DDR2, with an IC50 of 203 nM.

7rh (DDR1-IN-2) is a potent inhibitor of discoidin domain receptor 1 (DDR1), with an IC50 of 13.1 nM, and also less potently inhibits DDR2, with an IC50 of 203 nM.

Specifications

Chemical Name or Material 7rh
CAS 1429617-90-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H29F3N6O
Quantity 5 mg
Synonym DDR1-IN-2
Solubility Information DMSO : 62.5 mg/mL (114.35 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C(F)(F)F)=CC(CN2CCN(C)CC2)=C1)C3=CC=C(CC)C(C#CC4=CN5C(N=C4)=CC=N5)=C3
Molecular Weight (g/mol) 546.59
Formula Weight 546.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.