missing translation for 'onlineSavingsMsg'
Learn More

7H-Benz[de]anthracen-7-one, 99%, Thermo Scientific™

Product Code. 11476996
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476996 25 g 25g
11486996 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476996 Supplier Thermo Scientific Alfa Aesar Supplier No. B25322.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-05-3
Molecular Formula C17H10O
Molecular Weight (g/mol) 230.266
MDL Number MFCD00003585
InChI Key HUKPVYBUJRAUAG-UHFFFAOYSA-N
Synonym benzanthrone, 7h-benz de anthracen-7-one, benzanthrenone, ms-benzanthrone, dye, benzanthrone, benzoanthrone, mesobenzanthrone, 7h-benzo de anthracen-7-one, 1,9-benzanthrone
PubChem CID 6697
IUPAC Name benzo[a]phenalen-7-one
SMILES C1=CC=C2C(=C1)C3=CC=CC4=C3C(=CC=C4)C2=O

Specifications

Melting Point 171°C to 175°C
Quantity 25 g
Beilstein 1455646
Merck Index 14,1063
Formula Weight 230.27
Percent Purity 99%
Chemical Name or Material 7H-Benz[de]anthracen-7-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.