missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7beta-Hydroxy-5beta-cholanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 7beta-Hydroxy-5beta-cholanoic Acid |
| CAS | 10601-78-2 |
| Molecular Formula | C24 H40 O3 |
| Quantity | 100 mg |
| Synonym | 7beta-Hydroxy-5beta-cholan-24-oic Acid, 7beta-Hydroxy-5beta-cholanic Acid, 7beta-Hydroxy-5beta-cholanic Acid |
| InChI Formula | InChI=1S/C24H40O3/c1-15(7-10-21(26)27)17-8-9-18-22-19(11-13-24(17,18)3)23(2)12-5-4-6-16(23)14-20(22)25/h15-20,22,25H,4-14H2,1-3H3,(H,26,27)/t15?,16-,17-,18?,19+,20+,22?,23+,24+/m1/s1 |
| SMILES | CC(CCC(=O)O)[C@H]1CCC2C3[C@@H](O)C[C@H]4CCCC[C@]4(C)[C@H]3CC[C@@]12C |
| IUPAC Name | 4-[(5R,7S,9S,10S,13S,17R)-7-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
| Molecular Weight (g/mol) | 376.573 |
| Formula Weight | 376.298 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?