missing translation for 'onlineSavingsMsg'
Learn More

7Beta-Eplerenone, TRC

Product Code. 30388177
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388177 25 mg -
30371816 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388177 Supplier Toronto Research Chemicals Supplier No. TRCE58877025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209253-81-6
Molecular Formula C24H30O6
Molecular Weight (g/mol) 414.49
Synonym (7beta,11alpha,17alpha)-9,11-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic Acid gamma-Lactone Methyl Ester, Eplerenone EP Impurity E
SMILES COC(=O)[C@H]1CC2=CC(=O)CC[C@]2(C)[C@@]34O[C@@H]3C[C@@]5(C)[C@@H](CC[C@@]56CCC(=O)O6)[C@H]14

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H30O6/c1-21-7-4-14(25)10-13(21)11-15(20(27)28-3)19-16-5-8-23(9-6-18(26)30-23)22(16,2)12-17-24(19,21)29-17/h10,15-17,19H,4-9,11-12H2,1-3H3
Formula Weight 414.2042
Chemical Name or Material 7beta-Eplerenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.