missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7Alpha-Thiomethyl Spironolactone-d7 (Major), TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | 7alpha-Thiomethyl Spironolactone-d7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23H25D7O3S |
| Quantity | 1 mg |
| Synonym | 17α-Hydroxy-7α-(methylthio)-3-oxo-pregn-4-ene-21-carboxylic Acid γ-Lactone-d7, SC 26519-d7, 7α-Thiomethylspirolactone-d7, 7α-Thiomethyl-SL-d7 |
| SMILES | O=C1C([2H])([2H])C[C@@]2(C)C(C([2H])([2H])[C@@H](SC)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]45OC(C([2H])([2H])C5)=O)=C1[2H] |
| Molecular Weight (g/mol) | 395.61 |
| Percent Purity | >95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction