missing translation for 'onlineSavingsMsg'
Learn More

7Alpha-Hydroxyfrullanolide, TRC

Product Code. 30391817
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391817 10 mg -
30407874 25 mg -
30404450 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391817 Supplier Toronto Research Chemicals Supplier No. TRCH94270010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105578-87-8
Molecular Formula C15H20O3
Molecular Weight (g/mol) 248.32
Synonym 3-methylenenaphtho[1,2-b]furan-2(3H)-one, 7α-Hydroxy-4,11(13)-eudesmadien-12,6-olide, (3aR,5aR,9bS)-3a,4,5,5a,6,7,8,9b-Octahydro-3a-hydroxy-5a,9-dimethyl-3-methylene-naphtho[1,2-b]furan-2(3H)-one
IUPAC Name (3aR,5aR,9bS)-3a-hydroxy-5a,9-dimethyl-3-methylidene-4,5,6,7,8,9b-hexahydrobenzo[g][1]benzofuran-2-one
SMILES CC1=C2[C@@H]3OC(=O)C(=C)[C@]3(O)CC[C@@]2(C)CCC1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H20O3/c1-9-5-4-6-14(3)7-8-15(17)10(2)13(16)18-12(15)11(9)14/h12,17H,2,4-8H2,1,3H3/t12-,14+,15+/m0/s1
Formula Weight 248.1412
Chemical Name or Material 7Alpha-Hydroxyfrullanolide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.